Assay Details
Target Gene Details
Entrez Gene ID: | 8111 |
Gene Name: | G protein-coupled receptor 68 |
Gene Aliases: |
GPR12A, OGR1 |
Location: |
Chr.14:91232532-91264581 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR68 | NM_001177676.1 | 2 | 2605 | NP_001171147.1 |
| NM_003485.3 | 2 | 2559 | NP_003476.3 | |
| XM_005268110.4 | 2 | 3745 | XP_005268167.1 | |
| XM_005268111.3 | 2 | 2525 | XP_005268168.1 | |
| XM_005268112.3 | 2 | 2465 | XP_005268169.1 | |
| XM_006720262.3 | 2 | 2491 | XP_006720325.1 | |
| XM_011537196.2 | 3 | 2609 | XP_011535498.1 | |
| XM_011537197.2 | 2 | 2889 | XP_011535499.1 | |
| XM_011537198.2 | 2 | 3115 | XP_011535500.1 | |
| XM_011537199.2 | 2 | 2560 | XP_011535501.1 | |
| BC098567.1 | 2 | 2502 | AAH98567.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1046924 | Chr.14:90947458 - 91371146 on Build GRCh38 | Gain |
|
| nsv832857 | Chr.14:91185830 - 91325925 on Build GRCh38 | Loss |
|
| dgv141n21 | Chr.14:91175890 - 91233074 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs115978184,rs144110292] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map