Assay Details
Target Gene Details
Entrez Gene ID: | 9646 |
Gene Name: | CTR9 homolog, Paf1/RNA polymerase II complex component |
Gene Aliases: |
SH2BP1, TSBP, p150, p150TSP |
Location: |
Chr.11:10750987-10779755 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CTR9 | NM_014633.4 | 3 | 709 | NP_055448.1 |
| AK291794.1 | 3 | 445 | ||
| BC058914.1 | 3 | 456 | AAH58914.1 | |
| D63875.1 | 3 | 369 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483000 | Chr.11:10678454 - 12678453 on Build GRCh38 | Loss |
|
| nsv832063 | Chr.11:10596938 - 10763734 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map