Assay Details
Target Gene Details
Entrez Gene ID: | 7832 |
Gene Name: | BTG anti-proliferation factor 2 |
Gene Aliases: |
APRO1, PC3, TIS21 |
Location: |
Chr.1:203305536-203309602 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BTG2 | NM_006763.2 | 2 | 1487 | NP_006754.1 |
| BC105948.1 | AAI05949.1 | |||
| BC105949.1 | AAI05950.1 | |||
| U72649.1 | 2 | 1487 | AAB37580.1 |
More Information
Set Membership: |
|

Genomic Map