Assay Details
Target Gene Details
Entrez Gene ID: | 130872 |
Gene Name: | AHA1, activator of heat shock 90kDa protein ATPase homolog 2 (yeast) |
Gene Aliases: |
Hch1 |
Location: |
Chr.2:61177418-61191210 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AHSA2 | NM_001321305.1 | 6 | 2359 | NP_001308234.1 |
| XM_011532541.2 | 8 | 2711 | XP_011530843.1 | |
| XM_011532543.2 | 9 | 2995 | XP_011530845.1 | |
| XM_011532545.2 | 8 | 1419 | XP_011530847.1 | |
| XM_017003377.1 | 7 | 1317 | XP_016858866.1 | |
| XM_017003378.1 | 9 | 2670 | XP_016858867.1 | |
| AK091998.1 | 1 | 1891 |
Target Gene Details
Entrez Gene ID: | 9736 |
Gene Name: | ubiquitin specific peptidase 34 |
Gene Aliases: |
- |
Location: |
Chr.2:61187455-61470714 on Build GRCh38 |
Assay Gene Location: | Within Exon 80 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP34 | NM_014709.3 | 80 | 10596 | NP_055524.3 |
| AB011142.2 | 81 | 10651 | BAA25496.2 | |
| AB018272.1 | 23 | 3525 | BAA34449.1 | |
| AJ586138.1 | 78 | 10121 | CAE51938.1 | |
| AK001193.1 | 20 | 3187 | ||
| AK022637.1 | 10 | 1636 | ||
| AK023171.1 | 12 | 1873 | ||
| AL050092.1 | 15 | 2692 | CAB43264.1 | |
| AL050376.1 | 1 | 156 | ||
| AW293998.1 | ||||
| BC022783.1 | 17 | 2895 | AAH22783.1 | |
| BC062325.1 | 8 | 1350 | AAH62325.1 | |
| BC107761.1 | 7 | 1261 | AAI07762.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760616 | Chr.2:61136348 - 61200572 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map