Assay Details
Target Gene Details
Entrez Gene ID: | 23019 |
Gene Name: | CCR4-NOT transcription complex subunit 1 |
Gene Aliases: |
AD-005, CDC39, NOT1, NOT1H |
Location: |
Chr.16:58519946-58629886 on Build GRCh38 |
Assay Gene Location: | Within Exon 39 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNOT1 | NM_001265612.1 | 39 | 5889 | NP_001252541.1 |
| NM_016284.4 | 39 | 5904 | NP_057368.3 | |
| NR_049763.1 | 39 | 5889 | ||
| AB023224.2 | 26 | 3949 | BAA76851.2 | |
| AK056927.1 | ||||
| AK075125.1 | 9 | 1203 | ||
| AK300281.1 | ||||
| AK302495.1 | 16 | 2274 | ||
| AL117492.1 | 11 | 1743 | CAB55960.1 | |
| AL832769.1 | 39 | 5853 | ||
| AL833549.1 | 39 | 5863 | ||
| BC000779.2 | AAH00779.2 | |||
| BC024317.1 | 22 | 3303 | AAH24317.1 | |
| BX537840.1 | 39 | 5835 | CAD97851.1 | |
| CR749237.1 | 32 | 4844 | CAH18093.1 | |
| EF553522.1 | 39 | 5838 |
More Information
Set Membership: |
|

Genomic Map