Assay Details
Target Gene Details
Entrez Gene ID: | 8935 |
Gene Name: | src kinase associated phosphoprotein 2 |
Gene Aliases: |
PRAP, RA70, SAPS, SCAP2, SKAP-HOM, SKAP55R |
Location: |
Chr.7:26654772-26864743 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SKAP2 | NM_001303468.1 | 13 | 1884 | NP_001290397.1 |
| NM_003930.4 | 13 | 1992 | NP_003921.2 | |
| XM_017012771.1 | XP_016868260.1 | |||
| AF051323.1 | 13 | 1850 | AAC99296.1 | |
| BC002893.2 | 13 | 1836 | AAH02893.1 | |
| BC036044.1 | 13 | 1839 | AAH36044.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830929 | Chr.7:26548269 - 26710116 on Build GRCh38 | Gain |
|
| nsv1020378 | Chr.7:26640261 - 26694253 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map