Assay Details
Target Gene Details
Entrez Gene ID: | 79777 |
Gene Name: | acyl-CoA binding domain containing 4 |
Gene Aliases: |
HMFT0700 |
Location: |
Chr.17:45132600-45144181 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2666936 | Chr.17:45134343 - 45140631 on Build GRCh38 | Deletion |
|
| nsv953903 | Chr.17:45081533 - 45148633 on Build GRCh38 | Deletion |
|
| nsv527788 | Chr.17:44899824 - 45226241 on Build GRCh38 | Loss |
|
| nsv828022 | Chr.17:45137483 - 45160552 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map