Assay Details
Target Gene Details
Entrez Gene ID: | 27248 |
Gene Name: | endoplasmic reticulum lectin 1 |
Gene Aliases: |
C2orf30, CIM, CL24936, CL25084, HEL117, XTP3-B, XTP3TPB |
Location: |
Chr.2:53786931-53826928 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERLEC1 | NM_001127397.2 | NP_001120869.1 | ||
| NM_001127398.2 | NP_001120870.1 | |||
| NM_015701.4 | NP_056516.2 | |||
| XM_011532766.1 | XP_011531068.1 | |||
| AK001913.1 | BAA91974.1 | |||
| AK315477.1 | ||||
| AY358717.1 | AAQ89079.1 | |||
| AY453410.1 | AAR26725.1 | |||
| BC013129.1 | AAH13129.1 | |||
| BC022228.1 | AAH22228.1 | |||
| DB451941.1 | ||||
| EU794676.1 |
Target Gene Details
Entrez Gene ID: | 100302652 |
Gene Name: | GPR75-ASB3 readthrough |
Gene Aliases: |
- |
Location: |
Chr.2:53670293-53860033 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR75-ASB3 | NM_001164165.1 | NP_001157637.1 | ||
| DC314536.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834058 | Chr.2:53673537 - 53844874 on Build GRCh38 | Gain+Loss |
|
| nsv1009829 | Chr.2:53753425 - 53824790 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map