Assay Details
Target Gene Details
Entrez Gene ID: | 26019 |
Gene Name: | UPF2 regulator of nonsense transcripts homolog (yeast) |
Gene Aliases: |
HUPF2, RENT2, smg-3 |
Location: |
Chr.10:11920022-12043170 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UPF2 | NM_015542.3 | 19 | 3579 | NP_056357.1 |
| NM_080599.2 | 19 | 3749 | NP_542166.1 | |
| XM_017016034.1 | 18 | 3235 | XP_016871523.1 | |
| AB037829.1 | 18 | 3948 | ||
| AF301013.1 | 19 | 3572 | AAG33225.1 | |
| AF318574.1 | 19 | 3549 | AAG60689.1 | |
| AK000764.1 | 4 | 410 | BAA91369.1 | |
| AK074375.1 | 4 | 342 | BAB85064.1 | |
| AK096191.1 | BAC04721.1 | |||
| AK295209.1 | 6 | 648 | ||
| AL080198.1 | 13 | 1749 | CAB45771.1 | |
| AY013249.1 | 18 | 3498 | AAG48509.1 | |
| BC114964.1 | 19 | 3543 | ||
| BC115737.1 | 19 | 3543 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv955n54 | Chr.10:11936500 - 12034044 on Build GRCh38 | Loss |
|
| nsv831785 | Chr.10:11928851 - 12124076 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map