Assay Details
Target Gene Details
Entrez Gene ID: | 9507 |
Gene Name: | ADAM metallopeptidase with thrombospondin type 1 motif 4 |
Gene Aliases: |
ADAMTS-2, ADAMTS-4, ADMP-1 |
Location: |
Chr.1:161189725-161199080 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAMTS4 | NM_001320336.1 | 9 | 2583 | NP_001307265.1 |
| NM_005099.5 | 9 | 2744 | NP_005090.3 | |
| AB014588.1 | 9 | 2718 | ||
| AF148213.1 | 9 | 2724 | AAD41494.1 | |
| AK291573.1 | 9 | 2743 | ||
| AY358886.1 | 9 | 2736 | AAQ89245.1 | |
| BC063293.1 | 9 | 2743 | AAH63293.1 | |
| DQ364570.1 | 2 | 207 | ABC88384.1 |
More Information
Set Membership: |
|

Genomic Map