Assay Details
Target Gene Details
Entrez Gene ID: | 166979 |
Gene Name: | cell division cycle 20B |
Gene Aliases: |
G6VTS76519 |
Location: |
Chr.5:55112971-55173177 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CDC20B | NM_001145734.2 | 11 | 1985 | NP_001139206.2 |
| NM_001170402.1 | 12 | 2111 | NP_001163873.1 | |
| NM_152623.2 | 12 | 2099 | NP_689836.2 | |
| AK095246.1 | 12 | 2097 | BAC04508.1 | |
| BC136572.1 | 12 | 1981 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4833 | Chr.5:55082169 - 55126059 on Build GRCh38 | Deletion |
|
| nsv823075 | Chr.5:55113394 - 55114190 on Build GRCh38 | Loss |
|
| nsv830301 | Chr.5:54930046 - 55127677 on Build GRCh38 | Gain |
|
| nsv830302 | Chr.5:55107550 - 55259351 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map