Assay Details
Target Gene Details
Entrez Gene ID: | 4240 |
Gene Name: | milk fat globule-EGF factor 8 protein |
Gene Aliases: |
BA46, EDIL1, HMFG, HsT19888, MFG-E8, MFGM, OAcGD3S, SED1, SPAG10, hP47 |
Location: |
Chr.15:88898683-88913469 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MFGE8 | NM_001114614.2 | 7 | 1526 | NP_001108086.1 |
| NM_001310319.1 | 7 | 1550 | NP_001297248.1 | |
| NM_001310320.1 | 9 | 1735 | NP_001297249.1 | |
| NM_001310321.1 | 8 | 1794 | NP_001297250.1 | |
| NM_005928.3 | 8 | 1682 | NP_005919.2 | |
| AK095908.1 | 9 | 1645 | ||
| AK222735.1 | 8 | 1592 | BAD96455.1 | |
| AY141974.1 | 8 | 1601 | AAN08508.1 | |
| BC003610.2 | 7 | 1419 | AAH03610.1 | |
| BX537974.1 | 8 | 1556 | CAD97938.1 | |
| S56151.1 | 5 | 1025 | AAB19771.1 | |
| U58516.1 | 8 | 1592 | AAC50549.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1653 | Chr.15:88870409 - 88915341 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map