Assay Details
Target Gene Details
Entrez Gene ID: | 6760 |
Gene Name: | SS18, nBAF chromatin remodeling complex subunit |
Gene Aliases: |
SSXT, SYT |
Location: |
Chr.18:26016253-26091217 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893078 | Chr.18:25391998 - 26039471 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map