Assay Details
Target Gene Details
Entrez Gene ID: | 27166 |
Gene Name: | PRELI domain containing 1 |
Gene Aliases: |
CGI-106, PRELI, PX19, SBBI12 |
Location: |
Chr.5:177303762-177306959 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRELID1 | NM_001271828.1 | NP_001258757.1 | ||
| NM_013237.3 | NP_037369.1 | |||
| AF087858.1 | AAP97168.1 | |||
| AF112203.1 | AAF17191.1 | |||
| AF151864.1 | AAD34101.1 | |||
| AF153607.1 | AAD41089.1 | |||
| AF201925.1 | AAF09255.1 | |||
| AK312170.1 | ||||
| BC000007.2 | AAH00007.1 | |||
| BC007268.1 | AAH07268.1 | |||
| BC008307.1 | AAH08307.1 | |||
| BC008866.2 | AAH08866.1 | |||
| BC013733.1 | AAH13733.1 | |||
| BC013748.1 | AAH13748.1 | |||
| BG739785.1 | ||||
| BU956915.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv600364 | Chr.5:177299634 - 177432847 on Build GRCh38 | Loss |
|
| esv2759396 | Chr.5:177191316 - 177375443 on Build GRCh38 | Gain |
|
| nsv980764 | Chr.5:177304568 - 177306141 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map