Assay Details
Target Gene Details
Entrez Gene ID: | 51026 |
Gene Name: | golgi transport 1B |
Gene Aliases: |
CGI-141, GCT2, GOT1, GOT1B, YMR292W |
Location: |
Chr.12:21501765-21518403 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GOLT1B | NM_016072.4 | 5 | 900 | NP_057156.1 |
| AB097020.1 | 5 | 850 | BAC77373.1 | |
| AF068292.1 | AAF65181.1 | |||
| AF151899.1 | 5 | 834 | AAD34136.1 | |
| AK026999.1 | 1 | 82 | ||
| AK309736.1 | 4 | 780 | ||
| AK310889.1 | 6 | 1002 | ||
| AK311331.1 | 3 | 613 | ||
| AL136571.1 | 5 | 799 | CAB66506.1 | |
| AY358975.1 | 5 | 817 | AAQ89334.1 | |
| BC012455.1 | 5 | 821 | AAH12455.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv557741 | Chr.12:21425625 - 22016920 on Build GRCh38 | Gain |
|
| nsv1054220 | Chr.12:21459224 - 21539169 on Build GRCh38 | Gain |
|
| nsv528556 | Chr.12:21511945 - 21527429 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map