Assay Details
Target Gene Details
Entrez Gene ID: | 26515 |
Gene Name: | translocase of inner mitochondrial membrane 10 homolog B (yeast) |
Gene Aliases: |
FXC1, TIM10B, Tim9b |
Location: |
Chr.11:6481447-6484681 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TIMM10B | NM_012192.3 | 3 | 1302 | NP_036324.1 |
| AF150105.1 | 3 | 1291 | AAD40011.1 | |
| AF183415.1 | 3 | 1290 | AAG09684.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv972944 | Chr.11:6478192 - 6497815 on Build GRCh38 | Duplication |
|
| esv3625266 | Chr.11:6460034 - 6490367 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map