Assay Details
Target Gene Details
Entrez Gene ID: | 4352 |
Gene Name: | MPL proto-oncogene, thrombopoietin receptor |
Gene Aliases: |
C-MPL, CD110, MPLV, THCYT2, TPOR |
Location: |
Chr.1:43336875-43354464 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MPL | NM_005373.2 | NP_005364.1 | ||
| XM_017001320.1 | XP_016856809.1 | |||
| DQ234353.1 | ABB29982.1 | |||
| M90102.1 | AAA69971.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546128 | Chr.1:43301467 - 43451738 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map