Assay Details
Target Gene Details
Entrez Gene ID: | 102465485 |
Gene Name: | microRNA 6809 |
Gene Aliases: |
hsa-mir-6809 |
Location: |
Chr.2:217900513-217900628 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR6809 | NR_106867.1 |
Target Gene Details
Entrez Gene ID: | 7145 |
Gene Name: | tensin 1 |
Gene Aliases: |
MST091, MST122, MST127, MSTP091, MSTP122, MSTP127, MXRA6, PPP1R155, TNS |
Location: |
Chr.2:217799789-217978853 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Intron 6 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834541 | Chr.2:217838312 - 218052028 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map