Assay Details
Target Gene Details
Entrez Gene ID: | 9550 |
Gene Name: | ATPase H+ transporting V1 subunit G1 |
Gene Aliases: |
ATP6G, ATP6G1, ATP6GL, ATP6J, Vma10 |
Location: |
Chr.9:114587714-114598872 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP6V1G1 | NM_004888.3 | NP_004879.1 | ||
| AF038954.1 | AAC39868.1 | |||
| BC008452.1 | AAH08452.1 | |||
| BI596632.1 | ||||
| CR456971.1 | CAG33252.1 | |||
| CR542237.1 | CAG47033.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv948n27 | Chr.9:114193988 - 114693895 on Build GRCh38 | Gain |
|
| nsv831694 | Chr.9:114566360 - 114740505 on Build GRCh38 | Gain |
|
| dgv7706n100 | Chr.9:114182745 - 114697870 on Build GRCh38 | Gain |
|
| dgv33e196 | Chr.9:114030664 - 114604363 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map