Assay Details
Target Gene Details
Entrez Gene ID: | 25865 |
Gene Name: | protein kinase D2 |
Gene Aliases: |
HSPC187, PKD2, nPKC-D2 |
Location: |
Chr.19:46674316-46717127 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKD2 | NM_001079880.1 | 15 | 2327 | NP_001073349.1 |
| NM_001079881.1 | 15 | 2191 | NP_001073350.1 | |
| NM_001079882.1 | 13 | 1928 | NP_001073351.1 | |
| NM_016457.4 | 14 | 2606 | NP_057541.2 | |
| XM_005258716.2 | 13 | 1584 | XP_005258773.2 | |
| AF151021.1 | 2 | 132 | AAF36107.1 | |
| AF309082.1 | 14 | 1888 | AAK01149.1 | |
| AK074673.1 | 13 | 1567 | BAC11127.1 | |
| AK075266.1 | 13 | 1677 | BAC11508.1 | |
| AK092896.1 | 14 | 1978 | ||
| AK095884.1 | 15 | 1870 | ||
| AK300339.1 | 15 | 2141 | ||
| AK303629.1 | 13 | 1582 | ||
| AL050147.1 | 8 | 843 | CAB43292.1 | |
| BC015472.1 | 5 | 305 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833849 | Chr.19:46558831 - 46703060 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75185364] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map