Assay Details
Target Gene Details
Entrez Gene ID: | 23435 |
Gene Name: | TAR DNA binding protein |
Gene Aliases: |
ALS10, TDP-43 |
Location: |
Chr.1:11012622-11030528 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TARDBP | NM_007375.3 | NP_031401.1 | ||
| XM_017000863.1 | XP_016856352.1 | |||
| XM_017000864.1 | XP_016856353.1 | |||
| XM_017000865.1 | XP_016856354.1 | |||
| XM_017000866.1 | XP_016856355.1 | |||
| XM_017000867.1 | XP_016856356.1 | |||
| XM_017000868.1 | XP_016856357.1 | |||
| AK222754.1 | BAD96474.1 | |||
| AK295920.1 | ||||
| AK299454.1 | ||||
| AK312416.1 | ||||
| AL050265.1 | CAB43367.1 | |||
| BC071657.1 | AAH71657.1 | |||
| BC095435.1 | AAH95435.1 | |||
| BI464234.1 | ||||
| CD703666.1 | ||||
| CR533534.1 | CAG38565.1 | |||
| EF434181.1 | ||||
| EF434182.1 | ||||
| EF434183.1 | ||||
| HQ628633.1 | ||||
| HQ628634.1 | ||||
| HQ628635.1 | ||||
| HQ628636.1 | ||||
| HQ628637.1 | ||||
| U23731.1 | AAA70033.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832425 | Chr.1:10843788 - 11062918 on Build GRCh38 | Loss |
|
| nsv545407 | Chr.1:11000018 - 11377969 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map