Assay Details
Target Gene Details
Entrez Gene ID: | 79719 |
Gene Name: | alpha- and gamma-adaptin binding protein |
Gene Aliases: |
KPPP1, PPKP1, PPKP1A, p34 |
Location: |
Chr.15:67200675-67255198 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AAGAB | NM_001271885.1 | 10 | 2629 | NP_001258814.1 |
| NM_001271886.1 | 10 | 2890 | NP_001258815.1 | |
| NM_024666.4 | 10 | 2745 | NP_078942.3 | |
| XM_011522020.1 | 10 | 2995 | XP_011520322.1 | |
| AL136715.1 | 10 | 2655 | CAB66649.1 | |
| BQ185328.1 | 1 | 174 | ||
| CB243401.1 | 1 | 174 |
More Information
Set Membership: |
|

Genomic Map