Assay Details
Target Gene Details
Entrez Gene ID: | 688 |
Gene Name: | Kruppel like factor 5 |
Gene Aliases: |
BTEB2, CKLF, IKLF |
Location: |
Chr.13:73054976-73077542 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLF5 | NM_001286818.1 | 4 | 2192 | NP_001273747.1 |
| NM_001730.4 | 4 | 2806 | NP_001721.2 | |
| AF132818.1 | 4 | 2806 | AAF18307.1 | |
| AF287272.1 | 4 | 2581 | AAF88068.1 | |
| AK131397.1 | 1 | 2197 |
More Information
Set Membership: |
|

Genomic Map