Assay Details
Target Gene Details
Entrez Gene ID: | 90480 |
Gene Name: | GADD45G interacting protein 1 |
Gene Aliases: |
CKBBP2, CKbetaBP2, CRIF1, MRP-L59, PLINP, PLINP-1, PRG6, Plinp1 |
Location: |
Chr.19:12954156-12957254 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GADD45GIP1 | NM_052850.3 | 2 | 639 | NP_443082.2 |
| AF412816.1 | 2 | 596 | AAL55291.1 | |
| AF475095.1 | 2 | 597 | AAM10639.1 | |
| AF479749.1 | 2 | 621 | AAL85877.1 | |
| AI309741.1 | 1 | 134 | ||
| AJ295986.1 | 2 | 580 | CAC82499.1 | |
| AJ429498.1 | 2 | 597 | CAD22344.1 | |
| BC004944.1 | 2 | 585 | AAH04944.1 | |
| BC013027.2 | 2 | 594 | AAH13027.2 | |
| BC013039.2 | 2 | 594 | AAH13039.2 | |
| BC069200.1 | 2 | 599 | AAH69200.1 | |
| BI459923.1 | 2 | 644 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953977 | Chr.19:12666887 - 13095486 on Build GRCh38 | Deletion |
|
| nsv833754 | Chr.19:12876208 - 13028186 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map