Assay Details
Target Gene Details
Entrez Gene ID: | 8876 |
Gene Name: | vanin 1 |
Gene Aliases: |
HDLCQ8, Tiff66 |
Location: |
Chr.6:132680858-132714055 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VNN1 | NM_004666.2 | 7 | 2803 | NP_004657.2 |
| AJ132099.1 | 7 | 2797 | CAA10568.1 | |
| U39664.1 | 7 | 2798 | AAF21453.1 |
More Information
Set Membership: |
|

Genomic Map