Assay Details
Target Gene Details
Entrez Gene ID: | 2053 |
Gene Name: | epoxide hydrolase 2 |
Gene Aliases: |
CEH, SEH |
Location: |
Chr.8:27491002-27597559 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPHX2 | NM_001256482.1 | 3 | 444 | NP_001243411.1 |
| NM_001256483.1 | 2 | 397 | NP_001243412.1 | |
| NM_001256484.1 | 3 | 314 | NP_001243413.1 | |
| NM_001979.5 | 3 | 482 | NP_001970.2 | |
| XM_017013199.1 | 3 | 333 | XP_016868688.1 | |
| XM_017013200.1 | 3 | 333 | XP_016868689.1 | |
| AF233334.1 | 3 | 309 | AAG14966.1 | |
| AF233335.1 | 3 | 309 | AAG14967.1 | |
| AF233336.1 | 3 | 309 | AAG14968.1 | |
| AK096089.1 | 2 | 248 | ||
| AK096770.1 | 3 | 280 | ||
| AK294065.1 | 3 | 287 | ||
| AK303904.1 | 3 | 299 | ||
| AK308041.1 | 3 | 287 | ||
| AK310507.1 | 3 | 359 | ||
| BC007708.2 | 3 | 309 | AAH07708.1 | |
| BC011628.2 | 3 | 340 | AAH11628.1 | |
| BC013874.2 | 3 | 303 | AAH13874.1 | |
| BT006885.1 | 3 | 275 | AAP35531.1 | |
| CA397329.1 | 3 | 356 | ||
| DC331720.1 | 3 | 482 | ||
| L05779.1 | 3 | 316 | AAA02756.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831275 | Chr.8:27470773 - 27624088 on Build GRCh38 | Loss |
|
| nsv1020582 | Chr.8:27452251 - 27549327 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map