Assay Details
Target Gene Details
Entrez Gene ID: | 5529 |
Gene Name: | protein phosphatase 2 regulatory subunit B'epsilon |
Gene Aliases: |
- |
Location: |
Chr.14:63373892-63543361 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP2R5E | NM_001282179.1 | NP_001269108.1 | ||
| NM_001282180.1 | NP_001269109.1 | |||
| NM_001282181.1 | NP_001269110.1 | |||
| NM_001282182.1 | NP_001269111.1 | |||
| NM_006246.3 | NP_006237.1 | |||
| NR_104104.1 | ||||
| AB209811.1 | BAD93048.1 | |||
| AB451233.1 | ||||
| AB451358.1 | ||||
| AK299520.1 | ||||
| AK301796.1 | ||||
| AK313500.1 | ||||
| AK316430.1 | ||||
| BC048305.1 | AAH48305.1 | |||
| BC064358.1 | AAH64358.1 | |||
| BC092477.1 | AAH92477.1 | |||
| BC093766.1 | AAH93766.1 | |||
| BC101479.1 | ||||
| BC143231.1 | ||||
| BC143234.1 | ||||
| L76703.1 | AAB69752.1 | |||
| Z69029.1 | CAA93153.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv137n21 | Chr.14:62934068 - 63690399 on Build GRCh38 | Gain |
|
| esv2761522 | Chr.14:63298955 - 63400971 on Build GRCh38 | Gain |
|
| esv3634723 | Chr.14:63305196 - 63388109 on Build GRCh38 | Gain |
|
| dgv87e55 | Chr.14:63287645 - 63404698 on Build GRCh38 | Gain |
|
| nsv519574 | Chr.14:63301093 - 63391090 on Build GRCh38 | Gain |
|
| dgv88e55 | Chr.14:63312397 - 63388857 on Build GRCh38 | Gain |
|
| nsv832815 | Chr.14:63283340 - 63447182 on Build GRCh38 | Gain |
|
| dgv642e212 | Chr.14:63300256 - 63399207 on Build GRCh38 | Gain |
|
| nsv1051684 | Chr.14:63286198 - 63400959 on Build GRCh38 | Gain |
|
| dgv257n27 | Chr.14:63308966 - 63391090 on Build GRCh38 | Gain |
|
| dgv1925n100 | Chr.14:63298943 - 63400959 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map