Assay Details
Target Gene Details
Entrez Gene ID: | 6330 |
Gene Name: | sodium voltage-gated channel beta subunit 4 |
Gene Aliases: |
ATFB17, LQT10, Navbeta4 |
Location: |
Chr.11:118133377-118152915 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SCN4B | NM_001142348.1 | 3 | 972 | NP_001135820.1 |
| NM_001142349.1 | 4 | 1311 | NP_001135821.1 | |
| NM_174934.3 | 5 | 1374 | NP_777594.1 | |
| NR_024527.1 | 4 | 1157 | ||
| AK124886.1 | 3 | 766 | ||
| AY149967.1 | 5 | 1285 | AAN74584.1 | |
| BC035017.2 | 4 | 1275 | AAH35017.1 | |
| BX538328.1 | 2 | 672 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv556464 | Chr.11:118076983 - 118161527 on Build GRCh38 | Gain |
|
| esv3892103 | Chr.11:118081382 - 118161527 on Build GRCh38 | Gain |
|
| nsv1159938 | Chr.11:118075976 - 118157452 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map