Assay Details
Target Gene Details
Entrez Gene ID: | 26508 |
Gene Name: | hes related family bHLH transcription factor with YRPW motif-like |
Gene Aliases: |
HESR3, HEY3, HRT3, bHLHb33 |
Location: |
Chr.1:39623431-39639676 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HEYL | NM_014571.3 | 5 | 2002 | NP_055386.1 |
| XM_005270745.2 | 5 | 2056 | XP_005270802.1 | |
| BC006087.1 | 5 | 1972 | AAH06087.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv829649 | Chr.1:39505283 - 39638842 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map