Assay Details
Target Gene Details
Entrez Gene ID: | 10059 |
Gene Name: | dynamin 1 like |
Gene Aliases: |
DLP1, DRP1, DVLP, DYMPLE, EMPF, HDYNIV |
Location: |
Chr.12:32679200-32745650 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
More Information
Set Membership: |
|

Genomic Map