Assay Details
Target Gene Details
Entrez Gene ID: | 55197 |
Gene Name: | regulation of nuclear pre-mRNA domain containing 1A |
Gene Aliases: |
HsT3101, P15RS |
Location: |
Chr.18:35982385-36067594 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPRD1A | NM_001303411.1 | 8 | 2158 | NP_001290340.1 |
| NM_001303412.1 | NP_001290341.1 | |||
| NM_018170.4 | 7 | 2099 | NP_060640.2 | |
| XM_011526048.1 | XP_011524350.1 | |||
| XM_011526049.1 | 9 | 2238 | XP_011524351.1 | |
| XM_011526050.1 | 9 | 2347 | XP_011524352.1 | |
| AF419845.1 | 7 | 1916 | AAL15972.1 | |
| AK001518.1 | 7 | 2064 | BAA91736.1 | |
| BC000225.1 | AAH00225.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1065113 | Chr.18:35441391 - 36065348 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map