Assay Details
Target Gene Details
Entrez Gene ID: | 80127 |
Gene Name: | basal body orientation factor 1 |
Gene Aliases: |
C14orf45, CCDC176 |
Location: |
Chr.14:74019350-74078351 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3634932 | Chr.14:74053535 - 74063579 on Build GRCh38 | Loss |
|
| dgv1936n100 | Chr.14:73950133 - 74073431 on Build GRCh38 | Gain |
|
| dgv1938n100 | Chr.14:73984364 - 74075786 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map