Assay Details
Target Gene Details
Entrez Gene ID: | 146547 |
Gene Name: | protease, serine 36 |
Gene Aliases: |
- |
Location: |
Chr.16:31138926-31150094 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRSS36 | NM_001258290.1 | NP_001245219.1 | ||
| NM_001258291.1 | NP_001245220.1 | |||
| NM_173502.4 | NP_775773.2 | |||
| XM_011545758.2 | XP_011544060.1 | |||
| XM_017022968.1 | XP_016878457.1 | |||
| XM_017022969.1 | XP_016878458.1 | |||
| XM_017022970.1 | XP_016878459.1 | |||
| XM_017022971.1 | XP_016878460.1 | |||
| AJ627034.1 | CAF25303.1 | |||
| AK075142.1 | ||||
| AK290310.1 | ||||
| AK297997.1 | ||||
| AK301409.1 | ||||
| BC137396.1 | ||||
| BC144615.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv571820 | Chr.16:30892358 - 31180161 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map