Assay Details
Target Gene Details
Entrez Gene ID: | 9785 |
Gene Name: | DEAH-box helicase 38 |
Gene Aliases: |
DDX38, PRP16, PRPF16 |
Location: |
Chr.16:72093665-72112912 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHX38 | NM_014003.3 | NP_054722.2 | ||
| XM_005256269.1 | XP_005256326.1 | |||
| XM_011523484.1 | XP_011521786.1 | |||
| XM_011523485.1 | XP_011521787.1 | |||
| XM_017023913.1 | XP_016879402.1 | |||
| AK291634.1 | ||||
| AK293776.1 | ||||
| AK297269.1 | ||||
| AK297791.1 | ||||
| AK301074.1 | ||||
| AK309391.1 | ||||
| AK311169.1 | ||||
| BC004235.2 | AAH04235.1 | |||
| BC008340.1 | AAH08340.1 | |||
| BG719187.1 | ||||
| BM473438.1 | ||||
| D86977.1 |
Target Gene Details
Entrez Gene ID: | 54957 |
Gene Name: | thioredoxin like 4B |
Gene Aliases: |
DLP, Dim2 |
Location: |
Chr.16:72044261-72094316 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TXNL4B | NM_001142318.1 | NP_001135790.1 | ||
| CD517462.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428329 | Chr.16:71942441 - 72240448 on Build GRCh38 | Gain |
|
| esv2714683 | Chr.16:71950823 - 72151945 on Build GRCh38 | Deletion |
|
| nsv833273 | Chr.16:71937902 - 72118955 on Build GRCh38 | Loss |
|
| dgv3017n100 | Chr.16:72040933 - 72757730 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map