Assay Details
Target Gene Details
Entrez Gene ID: | 3645 |
Gene Name: | insulin receptor related receptor |
Gene Aliases: |
IRR |
Location: |
Chr.1:156840873-156858920 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| INSRR | NM_014215.2 | NP_055030.1 | ||
| J05046.1 | AAC31759.1 |
Target Gene Details
Entrez Gene ID: | 4914 |
Gene Name: | neurotrophic receptor tyrosine kinase 1 |
Gene Aliases: |
MTC, TRK, TRK1, TRKA, Trk-A, p140-TrkA |
Location: |
Chr.1:156815750-156881850 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NTRK1 | NM_001007792.1 | NP_001007793.1 | ||
| AK126428.1 | ||||
| CR936794.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831648 | Chr.1:156694085 - 156912126 on Build GRCh38 | Loss |
|
| nsv831659 | Chr.1:156771159 - 156980204 on Build GRCh38 | Loss |
|
| nsv998265 | Chr.1:156834329 - 156903671 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map