Assay Details
Target Gene Details
Entrez Gene ID: | 5291 |
Gene Name: | phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit beta |
Gene Aliases: |
P110BETA, PI3K, PI3KBETA, PIK3C1 |
Location: |
Chr.3:138652698-138834938 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIK3CB | NM_001256045.1 | 6 | 1106 | NP_001242974.1 |
| NM_006219.2 | 15 | 2240 | NP_006210.1 | |
| XM_005247530.2 | 17 | 2589 | XP_005247587.1 | |
| XM_006713659.3 | 16 | 2460 | XP_006713722.1 | |
| XM_011512895.2 | 18 | 2585 | XP_011511197.1 | |
| XM_011512896.1 | 13 | 1854 | XP_011511198.1 | |
| XM_017006619.1 | 16 | 2263 | XP_016862108.1 | |
| XM_017006620.1 | 15 | 2010 | XP_016862109.1 | |
| XM_017006621.1 | 14 | 1959 | XP_016862110.1 | |
| AK293754.1 | 6 | 1106 | ||
| AK302937.1 | 8 | 1119 | ||
| BC114432.1 | 15 | 2228 | AAI14433.1 | |
| CR749357.1 | 8 | 810 | CAH18210.1 | |
| S67334.1 | 15 | 2224 | AAB29081.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv591854 | Chr.3:138629239 - 139045387 on Build GRCh38 | Gain |
|
| nsv428422 | Chr.3:137344347 - 138686784 on Build GRCh38 | Gain+Loss |
|
| dgv4908n100 | Chr.3:138494324 - 139018846 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map