Assay Details
Target Gene Details
Entrez Gene ID: | 201501 |
Gene Name: | zinc finger and BTB domain containing 7C |
Gene Aliases: |
APM-1, APM1, ZBTB36, ZNF857C |
Location: |
Chr.18:48026886-48412639 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
More Information
Set Membership: |
|

Genomic Map