Assay Details
Target Gene Details
Entrez Gene ID: | 2737 |
Gene Name: | GLI family zinc finger 3 |
Gene Aliases: |
ACLS, GCPS, GLI3-190, GLI3FL, PAP-A, PAPA, PAPA1, PAPB, PHS, PPDIV |
Location: |
Chr.7:41960949-42237019 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLI3 | NM_000168.5 | 15 | 3609 | NP_000159.3 |
| XM_011515274.2 | 14 | 6233 | XP_011513576.1 | |
| XM_017011997.1 | 14 | 6266 | XP_016867486.1 | |
| AK299299.1 | 14 | 3495 | ||
| BC113616.1 | 14 | 3566 | ||
| BC117168.1 | 14 | 3566 | ||
| M57609.1 | 14 | 3572 | AAA52564.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1034175 | Chr.7:41864116 - 42018888 on Build GRCh38 | Gain |
|
| nsv5717 | Chr.7:41950387 - 41983735 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map