Assay Details
Target Gene Details
Entrez Gene ID: | 4685 |
Gene Name: | neural cell adhesion molecule 2 |
Gene Aliases: |
NCAM21 |
Location: |
Chr.21:20998315-21543324 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCAM2 | NM_004540.3 | NP_004531.2 | ||
| XM_011529576.2 | XP_011527878.1 | |||
| XM_011529579.2 | XP_011527881.1 | |||
| XM_017028353.1 | XP_016883842.1 | |||
| XM_017028354.1 | XP_016883843.1 | |||
| XM_017028355.1 | XP_016883844.1 | |||
| XM_017028356.1 | XP_016883845.1 | |||
| XM_017028357.1 | XP_016883846.1 | |||
| AK299054.1 | ||||
| BC052946.1 | AAH52946.1 | |||
| U75330.1 | AAB80803.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057774 | Chr.21:20958000 - 21000405 on Build GRCh38 | Gain |
|
| esv2751925 | Chr.21:18035494 - 21000411 on Build GRCh38 | Loss |
|
| nsv821690 | Chr.21:20770321 - 21203584 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map