Assay Details
Target Gene Details
Entrez Gene ID: | 229 |
Gene Name: | aldolase, fructose-bisphosphate B |
Gene Aliases: |
ALDB, ALDO2 |
Location: |
Chr.9:101420560-101435780 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALDOB | NM_000035.3 | 9 | 1490 | NP_000026.2 |
| AK026411.1 | 9 | 1041 | ||
| AK290795.1 | 9 | 1484 | ||
| AV645373.2 | 1 | 133 | ||
| AV656265.1 | 2 | 475 | ||
| BG616536.1 | 2 | 476 | ||
| X01098.1 | 9 | 1474 | CAA25572.1 | |
| X02747.1 | 9 | 1533 | CAA26526.1 |
More Information
Set Membership: |
|

Genomic Map