Assay Details
Target Gene Details
Entrez Gene ID: | 100 |
Gene Name: | adenosine deaminase |
Gene Aliases: |
- |
Location: |
Chr.20:44619519-44651758 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADA | NM_000022.3 | 11 | 1180 | NP_000013.2 |
| NM_001322050.1 | 10 | 1064 | NP_001308979.1 | |
| NM_001322051.1 | 10 | 1108 | NP_001308980.1 | |
| NR_136160.1 | 10 | 1115 | ||
| AK123988.1 | 1 | 1787 | ||
| AK223397.1 | 11 | 1130 | BAD97117.1 | |
| AL832305.1 | 6 | 1395 | ||
| AW450871.1 | 2 | 395 | ||
| BC007678.2 | 11 | 1141 | AAH07678.1 | |
| BC040226.1 | 11 | 1147 | AAH40226.1 | |
| CD014053.1 | 10 | 997 | ||
| CD014056.1 | 10 | 1048 | ||
| KJ534760.1 | 11 | 1029 | ||
| KU177892.1 | 11 | 1029 | ||
| KU177893.1 | 10 | 957 | ||
| KU177894.1 | 7 | 567 | ||
| KU177895.1 | 6 | 504 | ||
| X02994.1 | 11 | 1124 | CAA26734.1 |
More Information
Set Membership: |
|

Genomic Map