Assay Details
Target Gene Details
Entrez Gene ID: | 164153 |
Gene Name: | ubiquitin like 4B |
Gene Aliases: |
- |
Location: |
Chr.1:110112440-110113947 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBL4B | NM_203412.1 | 1 | 994 | NP_981957.1 |
| AA608970.1 | ||||
| AK098556.1 | 1 | 995 | BAC05331.1 | |
| BC058929.1 | 1 | 966 | AAH58929.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2343 | Chr.1:110089616 - 110125340 on Build GRCh38 | Insertion |
|
| nsv509424 | Chr.1:110104072 - 110134869 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map