Assay Details
Target Gene Details
Entrez Gene ID: | 3858 |
Gene Name: | keratin 10 |
Gene Aliases: |
BCIE, BIE, CK10, EHK, K10, KPP |
Location: |
Chr.17:40818117-40822621 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT10 | NM_000421.3 | NP_000412.3 | ||
| XM_005257343.3 | 7 | 1760 | XP_005257400.1 | |
| BC034697.1 | AAH34697.1 | |||
| J04029.1 | AAA60544.1 | |||
| M19156.1 | AAA59468.1 | |||
| M77663.1 | AAA59199.1 |
More Information
Set Membership: |
|

Genomic Map