Assay Details
Target Gene Details
Entrez Gene ID: | 9641 |
Gene Name: | inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase epsilon |
Gene Aliases: |
IKK-E, IKK-i, IKKE, IKKI |
Location: |
Chr.1:206470243-206496890 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IKBKE | NM_001193321.1 | NP_001180250.1 | ||
| NM_001193322.1 | NP_001180251.1 | |||
| NM_014002.3 | NP_054721.1 | |||
| XM_005273356.2 | XP_005273413.1 | |||
| XM_017002868.1 | XP_016858357.1 | |||
| AK093798.1 | ||||
| AK310737.1 | ||||
| AL706097.1 | ||||
| AY660028.1 | ||||
| BC105923.1 | AAI05924.1 | |||
| BC105924.1 | AAI05925.1 | |||
| BC107812.1 | AAI07813.1 | |||
| D63485.1 | ||||
| DA990116.1 | ||||
| DA999897.1 | ||||
| DB152295.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv564n100 | Chr.1:206468707 - 206488531 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map