Assay Details
Target Gene Details
Entrez Gene ID: | 23244 |
Gene Name: | PDS5 cohesin associated factor A |
Gene Aliases: |
PIG54, SCC-112, SCC112 |
Location: |
Chr.4:39822863-39977956 on Build GRCh38 |
Assay Gene Location: | Within Exon 34 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDS5A | NM_001100399.1 | 33 | 5843 | NP_001093869.1 |
| AB014548.1 | 21 | 3847 | BAA31623.1 | |
| AB621803.1 | 33 | 5857 | ||
| AF294791.1 | 33 | 5414 | AAM82347.1 | |
| AK001828.1 | 1 | 670 | ||
| BC009650.1 | 9 | 2428 | ||
| BC114218.1 | 33 | 5539 | AAI14219.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv594030 | Chr.4:39767966 - 39835508 on Build GRCh38 | Gain |
|
| nsv594031 | Chr.4:39808834 - 39856900 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map