Assay Details
Target Gene Details
Entrez Gene ID: | 4601 |
Gene Name: | MAX interactor 1, dimerization protein |
Gene Aliases: |
MAD2, MXD2, MXI, bHLHc11 |
Location: |
Chr.10:110207605-110287365 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MXI1 | NM_001008541.1 | NP_001008541.1 | ||
| NM_005962.4 | NP_005953.4 | |||
| NM_130439.3 | NP_569157.2 | |||
| AK303949.1 | ||||
| AK309718.1 | ||||
| AL538051.3 | ||||
| BC012907.1 | AAH12907.1 | |||
| BC035128.1 | ||||
| BG700036.1 | ||||
| BM927870.1 | ||||
| BX417051.2 | ||||
| CR536576.1 | CAG38813.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1112596 | Chr.10:110226242 - 110228242 on Build GRCh38 | Deletion |
|
| nsv951846 | Chr.10:110226043 - 110227342 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map