Assay Details
Target Gene Details
Entrez Gene ID: | 6284 |
Gene Name: | S100 calcium binding protein A13 |
Gene Aliases: |
- |
Location: |
Chr.1:153618799-153634092 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100A13 | NM_001024210.1 | 4 | 513 | NP_001019381.1 |
| NM_001024211.1 | 2 | 229 | NP_001019382.1 | |
| NM_001024212.1 | 2 | 200 | NP_001019383.1 | |
| NM_001024213.1 | 2 | 168 | NP_001019384.1 | |
| NM_005979.2 | 3 | 305 | NP_005970.1 | |
| XM_005245434.3 | 4 | 420 | XP_005245491.1 | |
| XM_011509862.2 | 6 | 720 | XP_011508164.1 | |
| XM_011509863.2 | 6 | 687 | XP_011508165.1 | |
| XM_011509864.1 | 4 | 510 | XP_011508166.1 | |
| XM_017002034.1 | 5 | 603 | XP_016857523.1 | |
| XM_017002035.1 | 5 | 604 | XP_016857524.1 | |
| XM_017002036.1 | 2 | 337 | XP_016857525.1 | |
| AA366085.1 | 2 | 124 | ||
| AK097132.1 | 4 | 1809 | ||
| AY987392.1 | AAX89402.1 | |||
| BC000632.2 | 1 | 78 | AAH00632.1 | |
| BC068064.1 | 1 | 64 | AAH68064.1 | |
| BC070291.1 | 3 | 305 | AAH70291.1 | |
| BM762088.1 | 2 | 202 | ||
| BT006724.1 | AAP35370.1 | |||
| BX472819.1 | 2 | 168 | ||
| CK817939.1 | 4 | 513 | ||
| CR542149.1 | CAG46946.1 | |||
| X99920.1 | 2 | 82 | CAA68188.1 |
More Information
Set Membership: |
|

Genomic Map