Assay Details
Target Gene Details
Entrez Gene ID: | 29082 |
Gene Name: | charged multivesicular body protein 4A |
Gene Aliases: |
C14orf123, CHMP4, CHMP4B, HSPC134, SHAX2, SNF7, SNF7-1, VPS32-1, VPS32A |
Location: |
Chr.14:24209581-24213830 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHMP4A | NM_014169.3 | NP_054888.2 | ||
| AB100262.2 | BAC79376.2 | |||
| AF161483.1 | AAF29098.1 | |||
| AF212243.1 | AAK14928.1 | |||
| AW732226.1 | ||||
| AY329084.1 | AAQ91193.1 | |||
| BC010893.1 | AAH10893.2 | |||
| BC107699.1 | AAI07700.1 | |||
| BC113533.1 | ||||
| BC113535.1 | ||||
| CB241867.1 |
More Information
Set Membership: |
|

Genomic Map