Assay Details
Target Gene Details
Entrez Gene ID: | 134728 |
Gene Name: | interleukin 1 receptor associated kinase 1 binding protein 1 |
Gene Aliases: |
AIP70, SIMPL |
Location: |
Chr.6:78867486-78979411 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IRAK1BP1 |
Target Gene Details
Entrez Gene ID: | 55023 |
Gene Name: | pleckstrin homology domain interacting protein |
Gene Aliases: |
BRWD2, DCAF14, WDR11, ndrp |
Location: |
Chr.6:78934419-79078294 on Build GRCh38 |
Assay Gene Location: | Within Exon 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHIP | NM_017934.5 | 31 | 3821 | NP_060404.3 |
| XM_005248729.4 | 31 | 3818 | XP_005248786.1 | |
| XM_011535918.2 | 28 | 3477 | XP_011534220.1 | |
| XM_017010989.1 | 33 | 3963 | XP_016866478.1 | |
| XM_017010990.1 | 30 | 3561 | XP_016866479.1 | |
| AF310250.1 | 8 | 828 | AAG45145.1 | |
| AJ303102.1 | 31 | 3608 | CAC83118.1 | |
| AK000712.1 | 4 | 451 | BAA91336.1 | |
| AL161957.1 | 9 | 884 | CAH10776.1 | |
| BC008909.2 | 6 | 549 | AAH08909.2 | |
| BC081569.1 | 9 | 864 | AAH81569.1 | |
| BC137488.1 | 31 | 3701 | ||
| DQ924532.1 | 31 | 3595 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830705 | Chr.6:78949580 - 79119813 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map