Assay Details
Target Gene Details
Entrez Gene ID: | 79443 |
Gene Name: | FYVE and coiled-coil domain containing 1 |
Gene Aliases: |
CATC2, CTRCT18, RUFY3, ZFYVE7 |
Location: |
Chr.3:45917899-45995824 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FYCO1 | NM_024513.3 | 18 | 5069 | NP_078789.2 |
| XM_006713333.3 | 19 | 6234 | XP_006713396.1 | |
| XM_006713334.3 | 18 | 6150 | XP_006713397.1 | |
| XM_011534111.2 | 19 | 5147 | XP_011532413.1 | |
| AJ292348.2 | 18 | 5053 | CAC33883.1 | |
| AK023397.1 | BAB14559.1 | |||
| AK074165.1 | 6 | 1221 | BAB84991.1 | |
| AL833308.2 | 11 | 3055 | CAD89924.1 | |
| BC007218.1 | AAH07218.1 | |||
| BX537863.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834678 | Chr.3:45762802 - 45968764 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map